1,1'-Dioctadecyl-3,3,3',3'-tetramethylindodicarbocyanine (DID) ≥95%, Ultrareine Qualität

Lieferant: AAT BIOQUEST

22034
AATB22034EA 120 EUR
AATB22034
1,1'-Dioctadecyl-3,3,3',3'-tetramethylindodicarbocyanine (DID) ≥95%, Ultrareine Qualität
1,1'-Dioctadecyl-3,3,3',3'-tetramethylindodicarbocyanine (DID)

DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.


  • Lipophilic
  • Even Cellular Staining


Bitte beachten For research use only.

Formel: C₆₁H₉₉ClN₂O₄
Molecular Weight: 959,923 g/mol
CAS-Nummer: 127274-91-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR