3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide] ≥95%

Lieferant: AAT BIOQUEST

Synonyme: DOC Iodide, 3-Ethyl-2-[3-[3-ethyl-2(3H)-benzoxazolylidene]-1-propenyl]benzoxazolium Iodide, 3,3'-Diethyloxacarbocyanine iodide, DiOC2(3) iodide, 3-Ethyl-2-(3-(3-ethylbenzo[d]oxazol-2(3H)-ylidene)prop-1-en-1-yl)benzo[d]oxazol-3-ium iodide

22038
AATB22038EA 120 EUR
AATB22038
3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide] ≥95%
3,3'-Diethyloxacarbocyanine iodide [DiOC2(3) iodide]

DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.


  • Lipophilic
  • Even Cellular Staining


Bitte beachten For research use only.

Formel: C₂₁H₂₁IN₂O₂
Molecular Weight: 460,32 g/mol
MDL: MFCD00011952
CAS-Nummer: 905-96-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR