3,3-Dipropyloxacarbocyanine iodide [DiOC3(3) iodide] ≥95%
Lieferant: AAT BIOQUEST
22039
AATB22039EA 120 EUR
AATB22039
3,3-Dipropyloxacarbocyanine iodide [DiOC3(3) iodide] ≥95%
3,3-Dipropyloxacarbocyanine iodide [DiOC3(3) iodide]
DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.
  • Lipophilic
  • Even Cellular Staining
Bitte beachten For research use only.