Tide Quencher™ 2 CPG 1000 Å, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ2 CPG

2204
AATB2204EA 273 EUR
AATB2204
Tide Quencher™ 2 CPG 1000 Å, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 2 CPG

TQ2 is designed to be a superior quencher to FAM, HEX, TET, JOE, TF2 and rhodamine 6G.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR