3,3-Dihexadecyloxacarbocyanine perchlorate [DiOC16(3) perchlorate]
Lieferant: AAT BIOQUEST
22042
AATB22042EA 120 EUR
AATB22042
3,3-Dihexadecyloxacarbocyanine perchlorate [DiOC16(3) perchlorate]
3,3-Dihexadecyloxacarbocyanine perchlorate [DiOC16(3) perchlorate]
DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.
  • Lipophilic
  • Even Cellular Staining
Bitte beachten For research use only.