1,1-Dihexadecyl-3,3,3,3-tetramethylindocarbocyanine perchlorate [DiIC16(3) perchlorate] ≥95%
Lieferant: AAT BIOQUEST
22044
AATB22044EA 120 EUR
AATB22044
1,1-Dihexadecyl-3,3,3,3-tetramethylindocarbocyanine perchlorate [DiIC16(3) perchlorate] ≥95%
1,1-Dihexadecyl-3,3,3,3-tetramethylindocarbocyanine perchlorate [DiIC16(3) perchlorate]
DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.
  • Lipophilic
  • Even Cellular Staining
Bitte beachten For research use only.
CAS-Nummer: 78566-75-3