DiOC₅(3) iodide ≥95%
Lieferant: AAT BIOQUEST
Synonyme:
3,3'-Dipentyloxacarbocyanine iodide, DiOC5(3)
22045
AATB22045EA
120
EUR
AATB22045
DiOC₅(3) iodide ≥95%
DiOC₅(3) iodide
DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.
- Lipophilic
- Even Cellular Staining
Bitte beachten For research use only.
Formel:
C₂₇H₃₃IN₂O₂ Molecular Weight: 544,47 g/mol |
CAS-Nummer:
53213-81-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...