3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] ≥95%

Lieferant: AAT BIOQUEST

Synonyme: 3-Hexyl-2-[3-(3-hexyl-1,3-benzoxazol-2(3H)-ylidene)-1-propen-1-yl]-1,3-benzoxazol-3-ium iodide, DiOC6(3) iodide, DiOC6(3), 3,3'-Dihexyloxacarbocyanine iodide, 3-Hexyl-2-[3-(3-hexyl-2(3H)benzoxazolylidene)-1-propenyl]benzoxazolium iodide

22046
AATB22046EA 120 EUR
AATB22046
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] ≥95%
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide]

DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.


  • Lipophilic
  • Even Cellular Staining


Bitte beachten For research use only.

Formel: C₂₉H₃₇IN₂O₂
Molecular Weight: 572.531 g/mol
MDL: MFCD00011954
CAS-Nummer: 53213-82-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR