1,1-Diododecyl-3,3,3,3-tetramethylindocarbocyanine-5,5-diSulphonic acid [DiIC12(3)-DS] ≥90%
Lieferant: AAT BIOQUEST
22050
AATB22050EA
152
EUR
AATB22050
1,1-Diododecyl-3,3,3,3-tetramethylindocarbocyanine-5,5-diSulphonic acid [DiIC12(3)-DS] ≥90%
1,1-Diododecyl-3,3,3,3-tetramethylindocarbocyanine-5,5-diSulphonic acid [DiIC12(3)-DS]
DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.
- Lipophilic
- Even Cellular Staining
Bitte beachten For research use only.