1,1-Diododecyl-3,3,3,3-tetramethylindodicarbocyanine-5,5-diSulphonic acid [DiIC12(5)-DS] ≥95%
Lieferant: AAT BIOQUEST
22051
AATB22051EA 152 EUR
AATB22051
1,1-Diododecyl-3,3,3,3-tetramethylindodicarbocyanine-5,5-diSulphonic acid [DiIC12(5)-DS] ≥95%
1,1-Diododecyl-3,3,3,3-tetramethylindodicarbocyanine-5,5-diSulphonic acid [DiIC12(5)-DS]
DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.
  • Lipophilic
  • Even Cellular Staining
Bitte beachten For research use only.