1,1-Dioctadecyl-3,3,3,3-tetramethylindodicarbocyanine-5,5-diSulphonic acid [DiIC18(5)-DS] ≥95%
Lieferant: AAT BIOQUEST
22054
AATB22054EA
152
EUR
AATB22054
1,1-Dioctadecyl-3,3,3,3-tetramethylindodicarbocyanine-5,5-diSulphonic acid [DiIC18(5)-DS] ≥95%
1,1-Dioctadecyl-3,3,3,3-tetramethylindodicarbocyanine-5,5-diSulphonic acid [DiIC18(5)-DS]
DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.
- Lipophilic
- Even Cellular Staining
Bitte beachten For research use only.