DiIC1(5) iodide ≥95%

Lieferant: AAT BIOQUEST

Synonyme: 1,1’,3,3,3’,3’-Hexamethylindodicarbocyanine iodide

22056
AATB22056EA 120 EUR
AATB22056
DiIC1(5) iodide ≥95%
DiIC1(5) iodide

DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.


  • Lipophilic
  • Even Cellular Staining


Bitte beachten For research use only.

Formel: C₂₇H₃₁IN₂
Molecular Weight: 510,5 g/mol
CAS-Nummer: 36536-22-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR