1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindotricarbocyanine iodide (DiR) ≥95%

Lieferant: AAT BIOQUEST

Synonyme: DiR iodide, 1,1-Dioctadecyl-3,3,3,3-tetramethylindotricarbocyanine iodide, 2-(7-(3,3-Dimethyl-1-octadecylindolin-2-ylidene)hepta-1,3,5-trien-1-yl)-3,3-dimethyl-1-octadecyl-3H-indol-1-ium iodide, DiIC18(7)

22070
AATB22070EA 120 EUR
AATB22070
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindotricarbocyanine iodide (DiR) ≥95%
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindotricarbocyanine iodide (DiR)

DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.


  • Lipophilic
  • Even Cellular Staining


Bitte beachten For research use only.

Formel: C₆₃H₁₀₁N₂I
Molecular Weight: 1013,39 g/mol
MDL: MFCD03427788
CAS-Nummer: 100068-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR