3,3'-Diethylthiacarbocyanine iodide [DiSC2(3)] ≥95%

Lieferant: AAT BIOQUEST

Synonyme: DiSC2(3), 3-Ethyl-2-[3-[3-ethyl-2(3H)-benzothiazolylidene]-1-propenyl]benzothiazolium Iodide, 3,3-Diethylthiacarbocyanine iodide, 3-Ethyl-2-(3-(3-ethylbenzo[d]thiazol-2(3H)-ylidene)prop-1-en-1-yl)benzo[d]thiazol-3-ium iodide

22073
AATB22073EA 152 EUR
AATB22073
3,3'-Diethylthiacarbocyanine iodide [DiSC2(3)] ≥95%
3,3'-Diethylthiacarbocyanine iodide [DiSC2(3)]

DiI, DiO, DiS, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.


  • Lipophilic
  • Even Cellular Staining


Bitte beachten For research use only.

Formel: C₂₁H₂₁IN₂S₂
Molecular Weight: 492.44 g/mol
MDL: MFCD00011956
CAS-Nummer: 905-97-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR