3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] ≥95%

Lieferant: AAT BIOQUEST

Synonyme: DiSC3(5), 3-Propyl-2-[5-[3-propyl-2(3H)-benzothiazolylidene]-1,3-pentadienyl]benzothiazolium iodide, 5-(3-Propylbenzothiazol-2-ylidene)-1-(3-propylbenzothiazolium-2-yl)-1,3-pentadiene odide, 3,3'-Dipropylthiadicarbocyanine iodide

22076
AATB22076EA 152 EUR
AATB22076
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)] ≥95%
3,3'-Dipropylthiadicarbocyanine iodide [DiSC3(5)]

DiI, DiO, DiS, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.


  • Lipophilic
  • Even Cellular Staining


Bitte beachten For research use only.

Formel: C₂₅H₂₇IN₂S₂
Molecular Weight: 546,53 g/mol
MDL: MFCD00075033
CAS-Nummer: 53213-94-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR