Tide Quencher™ 2 succinimidyl ester, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ2 SE

2210
AATB2210EA 307 EUR
AATB2210
Tide Quencher™ 2 succinimidyl ester, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 2 succinimidyl ester

TQ2 is designed to be a superior quencher to FAM, HEX, TET, JOE, TF2 and rhodamine 6G.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 479,55 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab86b1ad873b31c7e460abaadf45f2ddc53ab1f3&vid=f00b9569e86b3aeba9008814b9582a27d9f741c2&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR