1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate ≥95%

Lieferant: AAT BIOQUEST
Achtung

22102
AATB22102EA 120 EUR
AATB22102
1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate ≥95%
1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate

DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.

Formel: C₅₉H₉₇ClN₂O₄
MDL: MFCD00142354
CAS-Nummer: 41085-99-8
ADR: 5,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR