1,1-Dioctadecyl-3,3,3,3-tetramethylindocarbocyanine triflate (DiI triflate) ≥95%
Lieferant: AAT BIOQUEST
22103
AATB22103EA 307 EUR
AATB22103
1,1-Dioctadecyl-3,3,3,3-tetramethylindocarbocyanine triflate (DiI triflate) ≥95%
1,1-Dioctadecyl-3,3,3,3-tetramethylindocarbocyanine triflate (DiI triflate)
DiI, DiO, DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures.
  • Lipophilic
  • Even Cellular Staining
Bitte beachten For research use only.