Tide Quencher™ 2 alkyne, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ2 alkyne

2212
AATB2212EA 307 EUR
AATB2212
Tide Quencher™ 2 alkyne, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 2 alkyne

TQ2 is designed to be a superior quencher to FAM, HEX, TET, JOE, TF2 and rhodamine 6G.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 419,54 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f4eaaba435d9f3cfc6110f8db26f66a2893e089&vid=a5e54ca68e61e6a241465b6400bd8d238564ac8e&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR