C1-Bodi Fluor 500/510-C12 [equivalent to BODIPY® 500/510 C1, C12]
Lieferant: AAT BIOQUEST
22120
AATB22120EA
307
EUR
AATB22120
C1-Bodi Fluor 500/510-C12 [equivalent to BODIPY® 500/510 C1, C12]
Tests
Zelltests
The green fluorescent BODIPY FL C12 fatty acid can be used as a synthetic precursor to a wide variety of fluorescent phospholipids.
- Phospholipid Precursor
- pH-Insensitive Fluorescence
- Fluorescent In Aqueous and Lipids
Bitte beachten For research use only.