C1-Bodi Fluor 500/510-C12 [equivalent to BODIPY® 500/510 C1, C12]
Lieferant: AAT BIOQUEST
22120
AATB22120EA 307 EUR
AATB22120
C1-Bodi Fluor 500/510-C12 [equivalent to BODIPY® 500/510 C1, C12]
Tests Zelltests
The green fluorescent BODIPY FL C12 fatty acid can be used as a synthetic precursor to a wide variety of fluorescent phospholipids.
  • Phospholipid Precursor
  • pH-Insensitive Fluorescence
  • Fluorescent In Aqueous and Lipids
Bitte beachten For research use only.