Bodi Fluor FL-C12 [equivalent to BODIPY™ FL C12]
Lieferant: AAT BIOQUEST
22121
AATB22121EA
171
EUR
AATB22121
Bodi Fluor FL-C12 [equivalent to BODIPY™ FL C12]
Bodi Fluor FL-C12
Bodi Fluor FL-C12, a green fluorescent fatty acid, serves as a synthetic precursor for a wide array of fluorescent phospholipids.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only