5-(Iodoacetamido)fluorescein ≥75%

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 5-IAF

222
AATB222EA 307 EUR
AATB222
5-(Iodoacetamido)fluorescein ≥75%
5-(Iodoacetamido)fluorescein

5-IAF is one of the most popular green fluorescent dyes for thiol modifications of proteins along with fluorescein-5-maleimide.

Formel: C₂₂H₁₄INO₆
Molecular Weight: 515,25 g/mol
CAS-Nummer: 63368-54-7
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR