JC-10
Lieferant: AAT BIOQUEST
Synonyme: Enhanced JC-1
22204
AATB22204EA 152 EUR
AATB22204
JC-10
JC-10
JC-10 is capable of entering selectively into mitochondria, and changes reversibly its colour from green to orange as membrane potentials increase.
  • Improved water solubility
  • Low precipitation rates
  • Selective for mitochondria
  • Superior performance
When excited at 490 nm, the color of JC-10 changes reversibly from green to greenish orange as the mitochondrial membrane becomes more polarized. Both colors can be detected using the filters commonly mounted in all flow cytometers, so that green emission can be analysed in fluorescence channel 1 (FL1) and greenish orange emission in channel 2 (FL2)..
Bitte beachten For research use only.