Rhodamine 123 ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
3,6-Diamino-9-(2-(methoxycarbonyl)phenyl xanthylium chloride, 3,6-Diamino-9-(2-(methoxycarbonyl)phenyl)xanthylium chloride, 2-(6-Amino-3-imino-3H-xanthen-9-yl)benzoic acid methyl ester
22210
AATB22210EA
120
EUR
AATB22210
Rhodamine 123 ≥95%
Rhodamine 123
Positively charged rhodamine dyes (such as rhodamine esters and rosamines) are selectively localized in mitochondria, thus they are widely used for labeling mitochondria of live cells.
Formel:
C₂₁H₁₇ClN₂O₃ Molecular Weight: 380,82 g/mol |
MDL:
MFCD00012664 CAS-Nummer: 62669-70-9 ADR: 5,II |