Rhodamine B hexyl ester perchlorate
Lieferant: AAT BIOQUEST
22211
AATB22211EA
120
EUR
AATB22211
Rhodamine B hexyl ester perchlorate
Rhodamine B hexyl ester perchlorate
Positively charged rhodamine dyes (such as rhodamine esters and rosamines) are selectively localized in mitochondria, thus they are widely used for labeling mitochondria of live cells.
- Selective for mitochondria
- Easily-detectable signal
- Multiple applications
Bitte beachten For research use only.