Tide Quencher™ 3 amine, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ3 amine

2222
AATB2222EA 307 EUR
AATB2222
Tide Quencher™ 3 amine, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 3 amine

TQ3 is designed to be a superior quencher to TAMRA, TF3 and Cy3.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 609,62 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=646740da734386f7fc885022a26a863599fb8091&vid=40cf6a1ee8e0b203e7446a848d2c106091bb8182&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR