Tetramethylrhodamine ethyl ester perchlorate (TMRE) ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
Tetramethylrhodamine Ethyl Ester Perchlorate, TMRE, 3,6-bis(Dimethylamino)-9-[2-(ethoxycarbonyl)phenyl]-xanthylium perchlorate
22220
AATB22220EA
149
EUR
AATB22220
Tetramethylrhodamine ethyl ester perchlorate (TMRE) ≥95%
Tetramethylrhodamine ethyl ester perchlorate (TMRE)
Positively charged rhodamine dyes (such as rhodamine esters and rosamines) are selectively localized in mitochondria, thus they are widely used for labeling mitochondria of live cells.
Formel:
C₂₆H₂₇ClN₂O₇ Molecular Weight: 514,95 g/mol |
MDL:
MFCD00467973 CAS-Nummer: 115532-52-0 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...