DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide) ≥95%
Lieferant: AAT BIOQUEST
|
Achtung
|
22225
AATB22225EA
120
EUR
AATB22225
DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide) ≥95%
DASPEI (2-(4-(Dimethylamino)styryl)-N-ethylpyridinium iodide)
DASPEI is a styryl dye that stains mitochondria of live cells.
Formel:
C₁₇H₂₁IN₂ Molecular Weight: 380,273 g/mol |
MDL:
MFCD00011994 CAS-Nummer: 3785-01-1 ADR: 5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...