Tide Quencher™ 3 CPG 500 Å, Tide Quencher™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TQ3 CPG
2223
AATB2223EA
273
EUR
AATB2223
Tide Quencher™ 3 CPG 500 Å, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 3 CPG
TQ3 is designed to be a superior quencher to TAMRA, TF3 and Cy3.
- Stronger absorption
- Higher quenching efficiency
- Versatile reactive forms
- Excellent oligonucleotide and peptide labeling
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...