Tide Quencher™ 3WS acid, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ3WS acid

2227
AATB2227EA 307 EUR
AATB2227
Tide Quencher™ 3WS acid, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 3WS acid

TQ3WS is designed to be a superior quencher to TAMRA, TF3 and Cy3.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 912 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b82edf1fa09e62c999d409dc35679cea288d8ade&vid=cfb16d3273c20842f0f1ba84cb36e460a2fdf41d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR