DNA Red 600, CytoALL™
Lieferant: AAT BIOQUEST
22270
AATB22270EA 470 EUR
AATB22270
DNA Red 600, CytoALL™
DNA Red 600
CytoALL™ DNA Red 600 is a DNA-selective, cell permeable dye that stains all DNA contents including nuclei and mitochondria in live cells.
  • Stable for 24 months
It only requires a single step of incubating cells in the presence of CytoALL™ DNA Red 600.
Bitte beachten For Research Use Only