Tide Quencher™ 3 phosphoramidite, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ3 phosphoramidite

2228
AATB2228EA 307 EUR
AATB2228
Tide Quencher™ 3 phosphoramidite, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 3 phosphoramidite

TQ3 is designed to be a superior quencher to TAMRA, TF3 and Cy3.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 752,91 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR