Tide Quencher™ 3WS succinimidyl ester, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ3WS SE

2229
AATB2229EA 307 EUR
AATB2229
Tide Quencher™ 3WS succinimidyl ester, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 3WS succinimidyl ester

TQ3WS is designed to be a superior quencher to TAMRA, TF3 and Cy3.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 996,24 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b76f1b39331627c649091a4be3049cf59540a0a&vid=030c70a2451a19cb27a1ad629956758897c8099e&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR