Tide Quencher™ 3 succinimidyl ester, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ3 SE

2230
AATB2230EA 307 EUR
AATB2230
Tide Quencher™ 3 succinimidyl ester, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 3 succinimidyl ester

TQ3 is designed to be a superior quencher to TAMRA, TF3 and Cy3.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 550,59 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8032dc50edd575c2e0050fbec82ba210b8d3498b&vid=73b88e1ba503525fa1a6b4dea033819f7575de56&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR