Tide Quencher™ 3 azide, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ3 azide

2231
AATB2231EA 307 EUR
AATB2231
Tide Quencher™ 3 azide, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 3 azide

TQ3 is designed to be a superior quencher to TAMRA, TF3 and Cy3.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 521,59 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef6f82ae267c4f3243d8511ddea8233a4377aa48&vid=922509d8f3221e071a387c7e8d69c9eba72a524a&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR