Tide Quencher™ 3 alkyne, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ3 alkyne

2232
AATB2232EA 307 EUR
AATB2232
Tide Quencher™ 3 alkyne, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 3 alkyne

TQ3 is designed to be a superior quencher to TAMRA, TF3 and Cy3.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 490,58 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01d23d5bcb31f963f46beb63983715b88dcbd9fe&vid=64c76de16787f3b7c7b7922c11d186e6f55a1d45&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR