Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit
Lieferant: AAT BIOQUEST
22323
AATB22323EA 711 EUR
AATB22323
Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit
Tests Zelltests
It is sensitive and might be used for measuring active DNA synthesis at single-cell level.
  • Blue laser compatible
  • Highly reliable
  • Optimised for flow cytometry
Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit uses XdU which is incorporated into cellular DNA during DNA synthesis.
After fixing cells, the incorporated XdU is labelled with iFluor® 488 MTA. The resulted iFluor® 488-labeled DNA formed in cells is visualised in FITC Channel. Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry assay.
Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry assay.