3WS maleimide [TQ3 maleimide], Tide Quencher™
Lieferant: AAT Bioquest
2233
AATB2233EA 303 EUR
AATB2233
3WS maleimide [TQ3 maleimide], Tide Quencher™
3WS maleimide [TQ3 maleimide]
Tide Quencher™ 3WS (TQ3WS) has been developed as an advanced quenching agent, exhibiting superior properties over traditional quenchers such as TAMRA, TF3, and Cy3.
  • Stable for 12 months
  • Optimized for FRET applications
Quality tested in-house
Bitte beachten For Research Use Only