Tide Fluor™ 1 azide, Tide Fluor™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TF1 azide
2236
AATB2236EA
307
EUR
AATB2236
Tide Fluor™ 1 azide, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 1 azide
TF1 is designed to be a superior fluorophore alternative to EDANS.
- Stronger absorption and fluorescence intensity
- Lower environment-sensitive fluorescence
- pH-independent fluorescence from 3-11
- Improved sensitivity
Molecular Weight:
301,3 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...