Tide Fluor™ 1 alkyne, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF1 alkyne

2237
AATB2237EA 307 EUR
AATB2237
Tide Fluor™ 1 alkyne, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 1 alkyne

TF1 is designed to be a superior fluorophore alternative to EDANS.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 270,28 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c46976ca858f95c278675024edd213c76d971e3e&vid=fc6ce1fce69eb7bcb399843ab0c93f67f1f5bc78&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR