Tide Fluor™ 1 amine, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF1 amine

2239
AATB2239EA 307 EUR
AATB2239
Tide Fluor™ 1 amine, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 1 amine

TF1 is designed to be a superior fluorophore alternative to EDANS.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 503,35 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e04c509667c41689e7b4fd0a28dfbc0b42437c0&vid=890432fec76677e8ecb9578c49732a90a39d590e&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR