MycoLight™ Bacterial Viability Assay Kit
Lieferant: AAT BIOQUEST
22400
AATB22400EA 460 EUR
AATB22400
MycoLight™ Bacterial Viability Assay Kit
Tests Immunologische Tests
Mycolight™ Bacterial Viability Assay Kit provides two-colour fluorescence assay of bacterial viability in both gram-positive and negative bacterial cell.
  • Suitable for gram-positive and gram-negative bacteria
  • Contains all necessary components
  • Multiple applications
Bitte beachten For research use only.