MycoLight™ Ratiometric Bacterial Membrane Potential Kit *Red/Green Fluorescence*
Lieferant: AAT BIOQUEST
22401
AATB22401EA 460 EUR
AATB22401
MycoLight™ Ratiometric Bacterial Membrane Potential Kit *Red/Green Fluorescence*
Tests Immunologische Tests
AAT Bioquest's MycoLight™ Ratiometric Bacterial Membrane Potential Kit uses a fluorescent sensor that exhibits green fluorescence in both gram-positive and negative bacterial cells in low concentration, but the fluorescence shifts toward red emission at higher cytosolic concentrations due to the dye molecule aggregation caused by larger membrane potentials.
  • Suitable for gram-positive and gram-negative bacteria
  • Ratiometric detection
  • Contains all necessary Components
  • Multiple applications
Bitte beachten For research use only.