MycoLight™ Fluorimetric CTC Live Bacteria Quantification Kit
Lieferant: AAT BIOQUEST
22405
AATB22405EA 460 EUR
AATB22405
MycoLight™ Fluorimetric CTC Live Bacteria Quantification Kit
Tests Immunologische Tests
The CTC Flow Cytometric live bacteria assay kit provides an easy and convenient method for evaluating bacterial health and vitality as a function of the respiratory activity.
  • Suitable for gram-positive and gram-negative bacteria
  • Contains all necessary components
  • Multiple applications
  • Optimised for flow cytometry
Bitte beachten For research use only.