MycoLight™ Flow Cytometric Live Bacteria Assay Kit
Lieferant: AAT BIOQUEST
22407
AATB22407EA 460 EUR
AATB22407
MycoLight™ Flow Cytometric Live Bacteria Assay Kit
Tests Immunologische Tests
The MycoLight Flow cytometric live bacteria assay kit provides an easy and convenient method for evaluating bacterial vitality as a function of the intracellular esterase activity.
  • Suitable for gram-positive and gram-negative bacteria
  • Contains all necessary components
  • Multiple applications
  • Optimized for flow cytometry
Bitte beachten For research use only.