MycoLight™ Flow Cytometric Live Bacteria Assay Kit
Lieferant: AAT BIOQUEST
22407
AATB22407EA
460
EUR
AATB22407
MycoLight™ Flow Cytometric Live Bacteria Assay Kit
Tests
Immunologische Tests
The MycoLight Flow cytometric live bacteria assay kit provides an easy and convenient method for evaluating bacterial vitality as a function of the intracellular esterase activity.
- Suitable for gram-positive and gram-negative bacteria
- Contains all necessary components
- Multiple applications
- Optimized for flow cytometry
Bitte beachten For research use only.