MycoLight™ Live Bacteria Fluorescence Imaging Kit
Lieferant: AAT BIOQUEST
22409
AATB22409EA 460 EUR
AATB22409
MycoLight™ Live Bacteria Fluorescence Imaging Kit
Tests Immunologische Tests
The MycoLight™ Live Bacteria Fluorescence Imaging Kit provides an easy and convenient way for visualizing live bacteria through fluorescent microscope.
  • Suitable for gram-positive and gram-negative bacteria
  • Contains all necessary components
  • Multiple applications
  • Optimised for fluorescent microscopy
Bitte beachten For research use only.