MycoLight™ Rapid Fluorescence Bacterial Gram Stain Kit
Lieferant: AAT BIOQUEST
22413
AATB22413EA
614
EUR
AATB22413
MycoLight™ Rapid Fluorescence Bacterial Gram Stain Kit
Tests
Immunologische Tests
The MycoLight™ Rapid Fluorescence Bacterial Gram Stain Kit provides an easy and convenient way for determination of gram sign in live bacteria.
- Suitable for gram-positive and gram-negative bacteria
- Contains all necessary components
- Multiple applications
- Optimised for fluorescent microscopy
Bitte beachten For research use only.