MycoLight™ Rapid Fluorescence Gram-Positive Bacteria Staining Kit
Lieferant: AAT BIOQUEST
MycoLight™ Rapid Fluorescence Gram-Positive Bacteria Staining Kit provides a novel one-step fluorescence assay for the determination of gram sign in living bacteria.
- Suitable for gram-positive and gram-negative bacteria
- Contains all necessary components
- Multiple applications
- Optimized for fluorescent microscopy
Bitte beachten For research use only.