Tide Fluor™ 1 maleimide, Tide Fluor™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TF1 maleimide

2242
AATB2242EA 307 EUR
AATB2242
Tide Fluor™ 1 maleimide, Tide Fluor™ fluoreszierende Probe
Tide Fluor™ 1 maleimide

TF1 is designed to be a superior fluorophore alternative to EDANS.


  • Stronger absorption and fluorescence intensity
  • Lower environment-sensitive fluorescence
  • pH-independent fluorescence from 3-11
  • Improved sensitivity

Molecular Weight: 355,34 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3991d6e91aa031b21f5fcd95debdc8b2d04a7db1&vid=0440b934a679f58839b6c6ab89ff181ddd4d4c40&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR