Cell Navigator™ Flow Cytometric Exosome Staining Kit
Lieferant: AAT BIOQUEST
Designed for consistent and dependable results. The Cell Navigator™ Flow Cytometric Exosome Staining Kit addresses these challenges by enabling the detection of CD9-positive exosomes in purified or bead-bound samples.
- Rapid and accurate exosome detection in diverse sample matrices, including cell culture supernatants, biological fluids, and isolated exosome preparations
- Six-month shelf life
Exosomes, nanoscale (30 to 150 nm) membrane-bound vesicles secreted by cells, are central to intercellular communication.
Lieferumfang: Shipped with blue ice.
Bitte beachten To ensure effective detection and analysis, it is essential to enrich the cell culture media for exosomes before staining. Research use only.